Anti-IgA Mouse Monoclonal Antibody [clone: IL-A71]

Supplier: GeneTex
GTX43312
89275-736EA 581.75 USD
89275-736
Anti-IgA Mouse Monoclonal Antibody [clone: IL-A71]
Antibodies
Type: Secondary
Antigen: IgA
Clonality: Monoclonal
Clone: IL-A71
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR