Anti-DEFB103A Mouse Monoclonal Antibody [clone: L3-18b-E1]

GTX43299
89282-124EA 289.37 USD
89282-124
Anti-DEFB103A Mouse Monoclonal Antibody [clone: L3-18b-E1]
Mouse Monoclonal antibody [L3-18b-E1] to Defensin, beta-3 (a.a. 6-22)

Type: Primary
Antigen: DEFB103A
Clonality: Monoclonal
Clone: L3-18b-E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR