Anti-Dengue Types 1 2 3 4 Mouse Monoclonal Antibody [clone: BDI419]

Supplier: GeneTex
GTX43295
89299-804EA 609.34 USD
89299-804
Anti-Dengue Types 1 2 3 4 Mouse Monoclonal Antibody [clone: BDI419]
Antibodies
Type: Primary
Antigen: Dengue Types 1 2 3 4
Clonality: Monoclonal
Clone: BDI419
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR