Anti-IgL Goat Polyclonal Antibody

Supplier: GeneTex
GTX43289
89284-456EA 476.15 USD
89284-456
Anti-IgL Goat Polyclonal Antibody
Antibodies
HRP-conjugated Goat Polyclonal antibody to Human lambda Light chain

Recommended Starting Dilutions: For ELISA: Use at 1/4000 - 1/8000 Optimal dilutions should be determined experimentally by the researcher.

Type: Secondary
Antigen: IgL
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: Light Chain
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR