Anti-DSC3 Mouse Monoclonal Antibody [clone: Dsc3-U114]

Supplier: GeneTex
GTX43286
89333-998EA 429.65 USD
89333-998
Anti-DSC3 Mouse Monoclonal Antibody [clone: Dsc3-U114]
Antibodies
Mouse Monoclonal antibody [Dsc3-U114] to Desmocollin 3

Type: Primary
Antigen: DSC3
Clonality: Monoclonal
Clone: Dsc3-U114
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR