Anti-S-Tag Mouse Monoclonal Antibody [clone: SBSTAGa]

GTX43279
89281-174EA 565.83 USD
89281-174
Anti-S-Tag Mouse Monoclonal Antibody [clone: SBSTAGa]
Type: Primary
Antigen: S-Tag
Clonality: Monoclonal
Clone: SBSTAGa
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR