Anti-E. coli (SLT-2, STX-2) beta subunit Mouse Monoclonal Antibody [clone: 286-12212]

Supplier: GeneTex
GTX43249
89316-812EA 640.11 USD
89316-812
Anti-E. coli (SLT-2, STX-2) beta subunit Mouse Monoclonal Antibody [clone: 286-12212]
Antibodies
Type: Primary
Antigen: E. coli (SLT-2, STX-2) beta subunit
Clonality: Monoclonal
Clone: 286-12212
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR