Anti-VTCN1 Mouse Monoclonal Antibody (Phycoerythrin (PE)) [clone: MIH43]

Supplier: GeneTex
GTX43228
89274-840EA 468.94 USD
89274-840
Anti-VTCN1 Mouse Monoclonal Antibody (Phycoerythrin (PE)) [clone: MIH43]
Mouse monoclonal antibody [MIH43] to B7-H4

Type: Primary
Antigen: VTCN1
Clonality: Monoclonal
Clone: MIH43
Conjugation: Phycoerythrin (PE)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR