Anti-C3AR Mouse Monoclonal Antibody [clone: hC3aRZ1]

GTX43225
89274-838EA 451 USD
89274-838
Anti-C3AR Mouse Monoclonal Antibody [clone: hC3aRZ1]
Mouse monoclonal antibody [hC3aRZ1] to C3a Receptor

Type: Primary
Antigen: C3AR
Clonality: Monoclonal
Clone: hC3aRZ1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR