Anti-CD115 Rat Monoclonal Antibody [clone: 604B5 2E11]

Supplier: GeneTex
GTX43212
89276-028EA 551.18 USD
89276-028
Anti-CD115 Rat Monoclonal Antibody [clone: 604B5 2E11]
Antibodies
Rat monoclonal antibody [604B5 2E11] to M-CSF Receptor

Type: Primary
Antigen: CD115
Clonality: Monoclonal
Clone: 604B5 2E11
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR