Anti-SDC1 Mouse Monoclonal Antibody [clone: B-A38]

Supplier: GeneTex
GTX43203
89265-440EA 755.53 USD
89265-440
Anti-SDC1 Mouse Monoclonal Antibody [clone: B-A38]
Antibodies
Mouse monoclonal antibody [B-A38] to Syndecan-1

Recommended Starting Dilutions: Flow Cytometry: Neat - 1/5 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: SDC1
Clonality: Monoclonal
Clone: B-A38
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR