Anti-CD14 (CD14 Antigen) Mouse Monoclonal Antibody [clone: biG53]

Supplier: GeneTex
GTX43201
89294-410EA 688.47 USD
89294-410
Anti-CD14 (CD14 Antigen) Mouse Monoclonal Antibody [clone: biG53]
Antibodies
Mouse monoclonal antibody [biG53] to CD14

Type: Primary
Antigen: CD14 (CD14 Antigen)
Clonality: Monoclonal
Clone: biG53
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR