Anti-CD14 Mouse Monoclonal Antibody [clone: TK4]

Supplier: GeneTex
GTX43199
89274-834EA 659.64 USD
89274-834
Anti-CD14 Mouse Monoclonal Antibody [clone: TK4]
Antibodies
Mouse monoclonal antibody [TÜK4] to CD14

Recommended Starting Dilutions: Flow Cytometry: Neat - 1/10 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: TK4
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Cat Bovine, Dog, Goat, Mink Pig, Rabbit, Sheep, Cynomolgus MonkeyLlama
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR