Anti-CD14 Mouse Monoclonal Antibody [clone: TK4]

Supplier: GeneTex
GTX43198
89315-810EA 308.62 USD
89315-810
Anti-CD14 Mouse Monoclonal Antibody [clone: TK4]
Antibodies
Mouse monoclonal antibody [TÜK4] to CD14

Recommended Starting Dilutions: Flow Cytometry: Neat - 1/10 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: TK4
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Cat, Bovine, Dog, Goat, Mink, Pig, Rabbit, Sheep, Cynomolgus monkey, Llama
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR