Anti-CD147 Rat Monoclonal Antibody [clone: OX114]

GTX43195
89315-808EA 235.16 USD
89315-808
Anti-CD147 Rat Monoclonal Antibody [clone: OX114]
Rat monoclonal antibody [OX114] to CD147

Recommended Starting Dilutions: Flow Cytometry: Neat - 1/5 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD147
Clonality: Monoclonal
Clone: OX114
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR