Anti-CD151 Mouse Monoclonal Antibody [clone: 11G5a]

GTX43192
89315-804EA 234.96 USD
89315-804
Anti-CD151 Mouse Monoclonal Antibody [clone: 11G5a]
Mouse monoclonal antibody [11G5a] to CD151

Type: Primary
Antigen: CD151
Clonality: Monoclonal
Clone: 11G5a
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR