Anti-eltB Mouse Monoclonal Antibody [clone: B221M]

GTX43191
89265-704EA 473.92 USD
89265-704
Anti-eltB Mouse Monoclonal Antibody [clone: B221M]
Type: Primary
Antigen: eltB
Clonality: Monoclonal
Clone: B221M
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR