Anti-FASLG Hamster Monoclonal Antibody [clone: MFL4]

Supplier: GeneTex
GTX43174
89315-792EA 252.69 USD
89315-792
Anti-FASLG Hamster Monoclonal Antibody [clone: MFL4]
Hamster monoclonal antibody [MFL4] to Fas Ligand

Type: Primary
Antigen: Fasl
Clonality: Monoclonal
Clone: MFL4
Conjugation: Phycoerythrin (PE)
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR