Anti-CD182 (Chemokine (C-X-C motif)R 2) Mouse Monoclonal Antibody [clone: HC2]

GTX43171
89274-822EA 451 USD
89274-822
Anti-CD182 (Chemokine (C-X-C motif)R 2) Mouse Monoclonal Antibody [clone: HC2]
Mouse monoclonal antibody [HC2] to CXCR2

Type: Primary
Antigen: CD182 (Chemokine (C-X-C motif)R 2)
Clonality: Monoclonal
Clone: HC2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR