Anti-CD1W2 Mouse Monoclonal Antibody [clone: CC20]

Supplier: GeneTex
GTX43167
89265-432EA 832.32 USD
89265-432
Anti-CD1W2 Mouse Monoclonal Antibody [clone: CC20]
Antibodies
Type: Primary
Antigen: CD1W2
Clonality: Monoclonal
Clone: CC20
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bovine, Dog, Goat, Horse, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR