Anti-CD200 Mouse Monoclonal Antibody [clone: OX-104]

Supplier: GeneTex
GTX43165
89292-862EA 252.8 USD
89292-862
Anti-CD200 Mouse Monoclonal Antibody [clone: OX-104]
Mouse monoclonal antibody [OX-104] to CD200

Type: Primary
Antigen: CD200
Clonality: Monoclonal
Clone: OX-104
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR