Anti-CD200R (CD200R 1) Mouse Monoclonal Antibody [clone: OX108]

Supplier: GeneTex
GTX43160
89274-820EA 656.15 USD
89274-820
Anti-CD200R (CD200R 1) Mouse Monoclonal Antibody [clone: OX108]
Antibodies
Mouse monoclonal antibody [OX108] to CD200R

Recommended Starting Dilutions: Flow Cytometry: Neat - 1/10 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD200R (CD200R 1)
Clonality: Monoclonal
Clone: OX108
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR