Anti-CD209 Mouse Monoclonal Antibody [clone: MR-1]

Supplier: GeneTex
GTX43152
89274-818EA 655.6 USD
89274-818
Anti-CD209 Mouse Monoclonal Antibody [clone: MR-1]
Antibodies
Mouse monoclonal antibody [MR-1] to DC-SIGN

Recommended Starting Dilutions: Flow Cytometry: Neat - 1/10 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD209
Clonality: Monoclonal
Clone: MR-1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR