Anti-CR2 Mouse Monoclonal Antibody [clone: CC21]

Supplier: GeneTex
GTX43149
89280-278EA 558.63 USD
89280-278
Anti-CR2 Mouse Monoclonal Antibody [clone: CC21]
Antibodies
Type: Primary
Antigen: CR2
Clonality: Monoclonal
Clone: CC21
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR