Anti-CD229 Mouse Monoclonal Antibody [clone: HLy9.1.25]

GTX43142
89292-854EA 241.04 USD
89292-854
Anti-CD229 Mouse Monoclonal Antibody [clone: HLy9.1.25]
Mouse monoclonal antibody [HLy9.1.25] to CD229

Type: Primary
Antigen: CD229
Clonality: Monoclonal
Clone: HLy9.1.25
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR