Anti-IL2RA Mouse Monoclonal Antibody [clone: IL-A111]

Supplier: GeneTex
GTX43136
89276-370EA 858.81 USD
89276-370
Anti-IL2RA Mouse Monoclonal Antibody [clone: IL-A111]
Antibodies
Mouse monoclonal antibody [IL-A111] to IL2 Receptor alpha

Recommended Starting Dilutions: Flow Cytometry: Neat - 1/10 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: IL-A111
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bovine, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR