Anti-IL2RA Mouse Monoclonal Antibody [clone: OX-39]

Supplier: GeneTex
GTX43135
89276-020EA 693.6 USD
89276-020
Anti-IL2RA Mouse Monoclonal Antibody [clone: OX-39]
Antibodies
Mouse monoclonal antibody [OX-39] to IL2 Receptor alpha

Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: OX-39
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR