Anti-O157 Mouse Monoclonal Antibody [clone: B836M]

Supplier: GeneTex
GTX43111
89280-730EA 637.75 USD
89280-730
Anti-O157 Mouse Monoclonal Antibody [clone: B836M]
Antibodies
Type: Primary
Antigen: O157
Clonality: Monoclonal
Clone: B836M
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR