Anti-O157 Mouse Monoclonal Antibody [clone: B837M]

Supplier: GeneTex
GTX43110
89280-728EA 637.75 USD
89280-728
Anti-O157 Mouse Monoclonal Antibody [clone: B837M]
Antibodies
Type: Primary
Antigen: O157
Clonality: Monoclonal
Clone: B837M
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR