Anti-O157 Mouse Monoclonal Antibody [clone: B839M]

Supplier: GeneTex
GTX43106
89280-724EA 636.04 USD
89280-724
Anti-O157 Mouse Monoclonal Antibody [clone: B839M]
Antibodies
Type: Primary
Antigen: O157
Clonality: Monoclonal
Clone: B839M
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR