Anti-O157 Mouse Monoclonal Antibody [clone: BDI160]

Supplier: GeneTex
GTX43105
89280-722EA 655.47 USD
89280-722
Anti-O157 Mouse Monoclonal Antibody [clone: BDI160]
Antibodies
Type: Primary
Antigen: O157
Clonality: Monoclonal
Clone: BDI160
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR