Anti-CD4 Mouse Monoclonal Antibody [clone: 11354]

Supplier: GeneTex
GTX43093
89265-424EA 832.32 USD
89265-424
Anti-CD4 Mouse Monoclonal Antibody [clone: 11354]
Antibodies
Mouse monoclonal antibody [2-35] to CD4

Recommended Starting Dilutions: Flow Cytometry: Neat - 1/5 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 11354
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR