Anti-EBNA Mouse Monoclonal Antibody [clone: 211]

Supplier: GeneTex
GTX42991
89279-518EA 599.89 USD
89279-518
Anti-EBNA Mouse Monoclonal Antibody [clone: 211]
Type: Primary
Antigen: EBNA
Clonality: Monoclonal
Clone: 211
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR