Anti-DHEA Mouse Monoclonal Antibody [clone: 1-24.1 (BGN/1177/1241)]

Supplier: GeneTex
GTX41447
89279-378EA 632.43 USD
89279-378
Anti-DHEA Mouse Monoclonal Antibody [clone: 1-24.1 (BGN/1177/1241)]
Antibodies
Type: Primary
Antigen: DHEA
Clonality: Monoclonal
Clone: 1-24.1 (BGN/1177/1241)
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR