Anti-TCEB1 Chicken Polyclonal Antibody

Supplier: GeneTex
GTX41325
89275-496EA 518.72 USD
89275-496
Anti-TCEB1 Chicken Polyclonal Antibody
Chicken polyclonal antibody to TCEB1

Type: Primary
Antigen: TCEB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR