Anti-Epstein-Barr Virus Nuclear Antigen Mouse Monoclonal Antibody [clone: 0211 (6F9/60)]

GTX41277
89276-122EA 547.76 USD
89276-122
Anti-Epstein-Barr Virus Nuclear Antigen Mouse Monoclonal Antibody [clone: 0211 (6F9/60)]
Type: Primary
Antigen: Epstein-Barr Virus Nuclear Antigen
Clonality: Monoclonal
Clone: 0211 (6F9/60)
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR