Anti-Influenza A (Nucleoprotein) Mouse Monoclonal Antibody [clone: B249M]

Supplier: GeneTex
GTX41260
89278-592EA 662.9 USD
89278-592
Anti-Influenza A (Nucleoprotein) Mouse Monoclonal Antibody [clone: B249M]
Antibodies
Type: Primary
Antigen: Influenza A (Nucleoprotein)
Clonality: Monoclonal
Clone: B249M
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR