Anti-Influenza A (Nucleoprotein) Mouse Monoclonal Antibody [clone: B252M]

Supplier: GeneTex
GTX41258
89278-590EA 662.9 USD
89278-590
Anti-Influenza A (Nucleoprotein) Mouse Monoclonal Antibody [clone: B252M]
Antibodies
Type: Primary
Antigen: Influenza A (Nucleoprotein)
Clonality: Monoclonal
Clone: B252M
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR