Anti-Escherichia coli 015 Mouse Monoclonal Antibody [clone: 15/443]

Supplier: GeneTex
GTX41248
89280-558EA 647.62 USD
89280-558
Anti-Escherichia coli 015 Mouse Monoclonal Antibody [clone: 15/443]
Antibodies
Type: Primary
Antigen: Escherichia coli 015
Clonality: Monoclonal
Clone: 15/443
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: E. coli Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR