Anti-Escherichia coli 020 Mouse Monoclonal Antibody [clone: 6/332]

Supplier: GeneTex
GTX41242
89280-554EA 597.04 USD
89280-554
Anti-Escherichia coli 020 Mouse Monoclonal Antibody [clone: 6/332]
Antibodies
Type: Primary
Antigen: Escherichia coli 020
Clonality: Monoclonal
Clone: 6/332
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: E. coli Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR