Anti-Influenza A (Nucleoprotein) Mouse Monoclonal Antibody [clone: IVF8]

Supplier: GeneTex
GTX41238
89295-222EA 479.86 USD
89295-222
Anti-Influenza A (Nucleoprotein) Mouse Monoclonal Antibody [clone: IVF8]
Antibodies
Type: Primary
Antigen: Influenza A (Nucleoprotein)
Clonality: Monoclonal
Clone: IVF8
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR