Anti-Neisseria meningitidis Rabbit Polyclonal Antibody

GTX41219
89337-096EA 341.25 USD
89337-096
Anti-Neisseria meningitidis Rabbit Polyclonal Antibody
Type: Primary
Antigen: Neisseria meningitidis
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR