Anti-ESR2 Mouse Monoclonal Antibody [clone: 5/25]

Supplier: GeneTex
GTX41212
89262-502EA 1216.66 USD
89262-502
Anti-ESR2 Mouse Monoclonal Antibody [clone: 5/25]
Antibodies
Mouse monoclonal antibody [5/25] to Estrogen Receptor beta 5

Type: Primary
Antigen: ESR2
Clonality: Monoclonal
Clone: 5/25
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR