Anti-Influenza A Hemagglutinin H1 Mouse Monoclonal Antibody [clone: B217M]

Supplier: GeneTex
GTX41208
89288-260EA 617.51 USD
89288-260
Anti-Influenza A Hemagglutinin H1 Mouse Monoclonal Antibody [clone: B217M]
Antibodies
Type: Primary
Antigen: Influenza A Hemagglutinin H1
Clonality: Monoclonal
Clone: B217M
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR