Anti-CFP Mouse Monoclonal Antibody [clone: 39378]

Supplier: GeneTex
GTX41200
89262-494EA 765.79 USD
89262-494
Anti-CFP Mouse Monoclonal Antibody [clone: 39378]
Antibodies
Mouse monoclonal antibody [10-24] to Properdin

Type: Primary
Antigen: CFP
Clonality: Monoclonal
Clone: 39378
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR