Anti-F7 Mouse Monoclonal Antibody [clone: RFF-VII/2]

Supplier: GeneTex
GTX41194
89280-540EA 542.79 USD
89280-540
Anti-F7 Mouse Monoclonal Antibody [clone: RFF-VII/2]
Antibodies
Mouse monoclonal antibody [RFF-VII/2] to Factor VII

Type: Primary
Antigen: F7
Clonality: Monoclonal
Clone: RFF-VII/2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR