Anti-LcrV Mouse Monoclonal Antibody [clone: Va48]

Supplier: GeneTex
GTX41191
89336-650EA 759 USD
89336-650
Anti-LcrV Mouse Monoclonal Antibody [clone: Va48]
Antibodies
Type: Primary
Antigen: LcrV
Clonality: Monoclonal
Clone: Va48
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR