Anti-APOC1 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: GeneTex
GTX41185
89337-092EA 437.27 USD
89337-092
Anti-APOC1 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Goat polyclonal to Apolipoprotein C-I (HRP)

IA: 1/1000 - 1/3000. WB: 1/1000 - 1/3000. Predicted molecular weight: 9 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: APOC1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR