Anti-Neisseria gonorrhoeae Rabbit Polyclonal Antibody (Biotin)

Supplier: GeneTex
GTX41181
89337-090EA 659.34 USD
89337-090
Anti-Neisseria gonorrhoeae Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: Neisseria gonorrhoeae
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR