Anti-F8 Mouse Monoclonal Antibody [clone: RFF-VIIIC/8]

Supplier: GeneTex
GTX41177
89280-534EA 597.04 USD
89280-534
Anti-F8 Mouse Monoclonal Antibody [clone: RFF-VIIIC/8]
Antibodies
Mouse monoclonal antibody [RFF-VIIIC/8] to Factor VIII

Recommended Starting Dilutions:ELISA: 1/500 - 1/15000Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: F8
Clonality: Monoclonal
Clone: RFF-VIIIC/8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR